Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ziath DP5 Cube Rack Reader

Supplier: Azenta Life Sciences
Ziath
DP5-C-STD-80 DP5-CYS-C
77321-924EA 13893 USD InStock
77321-924 77321-926
Ziath DP5 Cube Rack Reader
Barcode Systems
A compact, versatile, quick and robust scanner with minimal cabling. The cube rack reader is a 2D datamatrix tube camera reader with a scan and decode time of 1 to 2 seconds. The cube can read all racks on the market including cryoboxes and SBS racks; even 384 racks.

  • Smallest 'scan all' scanner on the market
  • Camera based instrument resulting in rapid imaging and decoding
  • Automation friendly: footprint ideal for benchtop and integration
  • Minimal cabling
  • Includes masks for rack/cryobox compatibility
  • Easy to set up and easy to use; free DP5 software upgrades for life
  • DP5 Cube 2D scanner offers easy, out-of-the-box set up and is delivered pre-calibrated and ready to scan
  • Software includes remote control capability for integration applications

This instrument can scan all racks in the SBS format, regardless of the conguration or number of tubes, and by a quick and easy change of mask (included as standard) it can also scan all cryobox formats.

Our new automatic rack type detection feature means that you no longer have to configure the cube for all rack types. Simply place your rack on the scanning window and the software will determine rack type and run the scan - easy.

Compatible with 24, 48, 96, 240, 384 position SBS racks. Compatible with 81, 100, 196 position cryoboxes. Compatible with all commercially available datamatrix 2D coded tubes. Export data as CSV, Text, XLS, XML, JSON, Python, DB, PNG or E-mail. Operating system on host PC: Windows® 10, 11.

With Cryoprotection: Prevents the scanner window from misting when scanning a cold rack from a freeze. The passive technology does not use heat or blown air.

Certifications: CE marked.

Ordering information: Two-year warranty.

Packaging: One reader system per case.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty