Would you like to log in again?
Ziath DP5 Mirage Rack Reader
Supplier: Azenta Life Sciences
Ziath
DP5-UP-AT DP5-M-STD-80 DP5-CYS-M
77321-934EA 9038 USD InStock
77321-934 77321-930 77321-936
Ziath DP5 Mirage Rack Reader
Barcode Systems
Setting new standards for 2D barcode rack scanners. SBS only reader, easy to set up and easy to use.
  • The most compact camera based SBS 2D rack scanner available
  • Automation friendly: low profile allows use on robotic integrations for liquid handling systems
  • Innovative and unique scanning technology (patent pending)
  • Scans all current, commonly used, SBS format racks
  • Scans acoustic tube racks in seconds (i.e. AcoustiX)
  • Easy to set up and easy to use; free DP5 software upgrades for life
  • DP5 Mirage 2D scanner offers easy, out-of-the-box set up and is delivered pre-calibrated and ready to scan
  • Low profile and economic price makes the Mirage the perfect replacement for a traditional flat bed scanner
This instrument can scan all commonly used racks in the SBS format, and its low profile means that it can be easily added into liquid handling platforms and other automation systems.
Scans all currently available 24, 48, 96, 240 position racks. Export data as CSV, Text, XLS, XML, JSON, Python, DB, PNG or e-mail. Compatible with datamatrix 2D barcodes.
Scanning technology: CMOS camera
Scan time: approx 2 seconds
Operating system on host computer: Windows® 10, 11
With Cryoprotection: Prevents the scanner window from misting when scanning a cold rack from a freeze. The passive technology does not use heat or blown air.
Certifications: CE marked (reader and scanner).
Ordering information: Two-year warranty. Software includes remote control capability for integration applications.
Packaging: One reader system per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty