Would you like to log in again?
Ziath DataPaq™ Express Single Rack 2D Scanners
Supplier: Azenta Life Sciences
Ziath DataPaq™ Express
ZTS-1DR2-EXP ZTS-EXP-USB3-80
77321-948EA 12972 USD InStock
77321-948 77321-938
Ziath DataPaq™ Express Single Rack 2D Scanners
Barcode Systems
The Ziath DataPaq™ Express rack reader is a 2D datamatrix tube camera reader with an image capture time of only one second. DataPaq™ Express is marginally larger than the SBS rack itself, making it ideal for robotic integration.
  • Reads all 2D Datamatrix coded tubes
  • Export data to Excel, Text, XML, JSON and Database
  • Offers easy, out-of-the-box set up and is delivered pre-calibrated and ready to scan
  • With cryoprotection: Prevents the scanner window from misting when scanning a cold rack from a freeze
  • Passive technology does not use heat or blown air
  • Easy to set up and easy to use
  • Software includes remote control capability for integration applications
Compatible with all 2D barcoded tube racks in SBS format, the DataPaq™ Express is the smallest rack scanner currently available. Its camera based instrument delivers rapid imaging and decoding.
Operating system on host PC: Windows® 7, 8, 10.
Compatible with 24, 48, 96, 240, 384 SBS racks.
Certifications: CE marked.
Ordering information: Two-year warranty. One reader system per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty