Would you like to log in again?
Ziath DataPaq™ Mirage
Supplier: Azenta Life Sciences
Ziath DataPaq™
ZTS-MRG-80 ZTS-1DR2-MRG
77321-940EA 655 USD InStock
77321-940 77321-958
Ziath DataPaq™ Mirage
Barcode Systems
Economical camera-based reader with small footprint and low height, fitted with USB output and cryoprotection for use with cold racks.
  • Reads all 2D Datamatrix coded tubes
  • Low profile for easy integration with liquid handling and other automation platforms
  • Export data to Excel, Text, XML, JSON and image files
  • Easy to set up and easy to use
  • Offers easy, out-of-the-box set up and is delivered pre-calibrated and ready to scan
  • With Cryoprotection: Prevents the scanner window from misting when scanning a cold rack from a freeze
  • Our passive technology does not use heat or blown air
Software includes remote control capability for integration applications.
Operating system on host PC: Windows® 7, 8, 10.
Compatible with 24, 48, 96, 240, 384 SBS racks.
Certifications: CE marked.
Ordering information: Two-year warranty.
Packaging: One reader system per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty