Would you like to log in again?
Ziath DataPaq™ Express-NMR Rack Reader
Supplier: Azenta Life Sciences
Ziath DataPaq™
ZTS-EXP-NMR
77321-946EA 22097 USD InStock
77321-946
Ziath DataPaq™ Express-NMR Rack Reader
Barcode Systems
Express NMR tube reader can read all 2D-barcoded NMR tubes including Bruker™ and Norell™ and is supplied with a handy slide-out tray designed to hold Bruker tube racks in the correct orientation and in the correct position for successful barcode reading.
  • Capable of reading all 2D-barcoded NMR tube caps
  • Inverted design especially for reading NMR tubes
  • Export data to Excel, Text, XML, JSON and Database
  • Easy to set up and easy to use
  • Offers easy, out-of-the-box set up and is delivered pre-calibrated and ready to scan
  • Industry-leading fast read and decode time for a full rack of under 3 seconds
Software includes remote control capability for integration applications.
Operating system on host PC: Windows® 10, 11.
Certifications: CE marked.
Ordering information: Two-year warranty. Includes DataPaq™ Express instrument with separate controller module, mounting housing for inverted Express optical module, precision engineered slide and drawer with dedicated rack holder, power adaptor and all necessary connecting cables; one reader.
Packaging: One reader system per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty