Would you like to log in again?
Ziath Handheld 2 Single Tube 2D Barcode Reader
Supplier: Azenta Life Sciences
Ziath
ZTS-HND2
77321-950EA 3251 USD InStock
77321-950
Ziath Handheld 2 Single Tube 2D Barcode Reader
Barcode Systems
The Ziath Handheld 2 reader is a truly portable reader which can be held and operated in one hand; leaving the other free for tube handling. Requires no external computer.
  • Compatible with all 2D datamatrix coded tubes
  • Portable, independent and compact
  • Pre-loaded software boots up immediately
  • Battery life of up to a week with casual use
The first truly portable reader that allows sample management on the move. The Handheld is light and robust with an intuitive user interface. It has five operating modes; simple tube reading, read and display data, picklist operations, edit data/ add new sample, and the optional 'Scripts', which enables wireless access to your database. Connection to a database, Excel, FreezerPro, LIMS or wireless keyboard wedge are all options.
Certifications: CE marked.
Packaging: One reader system per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty