Would you like to log in again?
Ziath UNO and DUO Single Tube Reader
Supplier: Azenta Life Sciences
Ziath
ZTS-DUO-80 ZTS-UNO-80 ZTS-UP-AT
77321-944EA 1840 USD InStock
77321-944 77321-960 77321-962
Ziath UNO and DUO Single Tube Reader
Barcode Systems
Uno and Duo single-tube readers scan all 2D datamatrix barcoded tubes. Simply hold the tube to the large scanner window and the code is instantly displayed or added directly into your application.
  • Reads all 2D Datamatrix coded tubes, including Azenta Acoustic tubes (with Acoustic add-on)
  • Uno features higher resolution camera that can often decode older, badly-printed barcodes
  • Duo combines the Datamatrix reader with a linear barcode scanner to read rack codes or 1D barcodes along the edge of some tubes
  • Super-fast electronics capable of reading 2D barcodes from any currently available tube on the market, including 384-well tubes
  • Large scan windows make it easy to present the tubes
  • Easy to set up and easy to use
  • Offers easy, out-of-the-box set up and is delivered pre-calibrated and ready to scan
The Uno single tube reader features a higher resolution camera than the previous model that can often decode older, badly-printed barcodes more easily. The Duo reader combines the Datamatrix reader with a linear barcode scanner to read rack codes or 1D barcodes along the edge of some tubes.
These rugged new tube readers feature a high-resolution camera with super-fast electronics capable of reading 2D barcodes from any currently available tube on the market including 384-well tubes.
Operating system on host PC: Windows® 10, 11.
With cryoprotection: Prevents the scanner window from misting when scanning a cold rack from a freeze. The passive technology does not use heat or blown air.
Certifications: CE marked (readers).
Ordering information: Two-year warranty.
Packaging: One reader system per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty