Would you like to log in again?
Cap2™ PCR Tubes, 0.2 ml Dual-Cap Sample Collection
Supplier: Azenta Life Sciences
Cap2™
4TI-0791
77321-968CS 910 USD InStock
77321-968
Cap2™ PCR Tubes, 0.2 ml Dual-Cap Sample Collection
Tubes PCR Tubes
A unique PCR tube featuring a patent pending dual-cap mechanism, 2D datamatrix code as well as a human readable identification, allowing for wide-ranging workflow versatility. Innovative features allow for utilization as a consumable suitable for sample collection/transport that can be directly processed for analysis, ideal for genetic testing workflows.
  • Patent pending dual-cap design with both hinged lid and screw cap
  • Sample identification from 2D code and human readable number
  • Tab on side of cap includes a 2D datamatrix code and human readable number
  • Tab on collar of hinged lid makes for quick and secure sample tracking
  • Capability to work with automation, tubes are compatible with 48-format SBS footprint rack
  • PCR tube with patent pending cap-in-cap design, sample identifiers and automation friendly capabilities offer a flexible, scalable solution
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Leak tested
  • Polypropylene, ultra-thin walled, low DNA binding tubes make for optimum PCR results and mitigation against DNA loss from samples
  • 0.2 ml (200 µl) total well capacity
The novel cap-in-cap design features a hinged lid and automation friendly screw cap, offering flexibility for use in manual workflows, alongside mitigating against contamination, as well as workflow scale up capabilities and consumable consolidation. This unique solution to error-prone, slow and labor-intensive workflows offers a flexible, scalable solution for PCR and genetic testing applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty