Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brady® HH83 Handheld Barcode Scanner and Mobile Computer

Supplier: Brady
Brady®
310378
77322-100EA 2359.78 USD InStock
77322-100
Brady® HH83 Handheld Barcode Scanner and Mobile Computer
Barcode Systems
Use this high-performance tool for all your mobile computing and data collection needs. Quick and accurate, this ruggedized scanner is designed for all-day use.

  • Android 10 operating system
  • RFID capabilities: NFC/HF

Reader is ruggedized and built for data collection in challenging environments. Ergonomic design for extended usage. IP 65 rated - protects against dust and moisture; suitable for moderate outdoor use. Five-foot drop test protection safeguards against damages. Get best-in-class RFID read performance with the software-controllable adaptive cross dipole antenna. Battery hot swap: replace an empty battery and install a fully charged one in seconds.

Device indicator: RGB LED
vibration for haptic feedback
Frequency: ETSI 865.6 to 867.6 MHz or FCC/IC 902 to 928 MHz

Delivery information: Has lithium-ion battery; must ship ground. Includes (1) HH83 handheld barcode scanner and mobile computer.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty