Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brady® HH83 Handheld RFID Reader - UHF, NFC, Barcode

Supplier: Brady
Brady®
310382
77322-102EA 4299.94 USD InStock
77322-102
Brady® HH83 Handheld RFID Reader - UHF, NFC, Barcode
Barcode Systems
Get high-performance RFID and barcode scanning with this HH83 Handheld RFID Reader. Ruggedized and ergonomic, it is built for daily scanning in the most challenging industrial environments.

  • Android 10 operating system
  • RFID capabilities: NFC/HF and UHF
  • Reading speed up to 1000 tags per second

A ruggedized scanner for data collection in challenging industrial environments. Ergonomic design for extended usage. IP 65 rated – protects against dust and moisture; suitable for moderate outdoor use. Five-foot drop test protection safeguards against damages. Get best-in-class RFID read performance with the software-controllable adaptive cross dipole antenna . Battery hot swap: replace an empty battery and install a fully charged one in seconds.

Device indicator: RGB LED
vibration for haptic feedback.
Frequency: ETSI 865.6 to 867.6 MHz or FCC/IC 902 to 928 MHz

Delivery information: Has lithium-ion battery; must ship ground. Includes handheld device; changeable battery (pre-installed); wrist strap; safety and regulations card.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty