Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brady® HH85 Handheld RFID Reader with Pistol Grip - UHF, NFC, Barcode

Supplier: Brady
Brady®
310385
77322-104EA 5066.98 USD InStock
77322-104
Brady® HH85 Handheld RFID Reader with Pistol Grip - UHF, NFC, Barcode
Barcode Systems
Get high-performance RFID and barcode scanning with this HH85 Handheld RFID Reader. Ruggedized and ergonomic, it’s built for daily scanning in the most challenging industrial environments. A pistol grip and additional battery life are ideal for extended use.

  • Android 10 operating system
  • RFID capabilities: NFC/HF and UHF
  • Reading speed up to 1000 tags per second

Ruggedized for industrial environments. Designed for longer use with an ergonomic pistol grip and extended battery life. IP 65 rated - protects against dust and moisture, suitable for moderate outdoor use. Five-foot drop test protection safeguards against damages. Get best-in-class RFID read performance with the software-controllable adaptive cross dipole antenna . Battery hot swap: replace an empty battery and install a fully charged one in a matter of seconds.

Device indicator: RGB LED
vibration for haptic feedback
Frequency: ETSI 865.6 to 867.6 MHz or FCC/IC 902 to 928 MHz

Delivery information: Has lithium-ion battery; must ship ground. Includes handheld device; changeable battery (pre-installed); wrist strap; safety and regulations card.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty