Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brady® FR22 Fixed UHF RFID Reader

Supplier: Brady
Brady®
311938
77322-128EA 3383.98 USD InStock
77322-128
Brady® FR22 Fixed UHF RFID Reader
Barcode Systems
Offering unparalleled flexibility and connectivity, the FR22 Fixed RFID reader enables automated data collection and exchange between a variety of platforms and use cases.

  • Linux operating system
  • RFID capabilities: UHF
  • Reading speed up to 1000 tags per second

Features integrated Bluetooth, Wi-Fi, LAN, USB, and GPIO, as well as HDMI video output. Linux OS provides edge computing capabilities, allowing users to run plugins or applications directly on the FR22 reader. Plug and play support for accessories including a 16-port antenna multiplexor and beam forming antenna with 28 individual beams for directionality detection.

Device indicator: 4 LEDs indicating the enabled antennas
Connection LED
Application LED
Power LED
RF LED
Extension port LED
External antenna: 4 ports RP-SMA female
Frequency: ETSI 865.6 to 867.6 MHz or FCC/IC 902 to 928 MHz

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty