Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brady® FR22 Fixed RFID Reader with BFA915 RFID Antenna Kit

Supplier: Brady
Brady®
311944
77322-136EA 5849.06 USD InStock
77322-136
Brady® FR22 Fixed RFID Reader with BFA915 RFID Antenna Kit
Barcode Systems
Get advanced RFID reading coverage and accuracy with the FR22 Fixed RFID Reader and BFA (Beam Forming Antenna) kit.

  • Linux operating system
  • RFID capabilities: UHF
  • Reading speed up to 1000 tags per second

Kit includes FR22 Fixed RFID Reader and BFA (beam forming antenna) antenna.

Device indicator: Buzzer for sound indications
4 LEDs indicating the enabled antennas
Connection LED
Application LED
Extension port LED
Power LED
RF LED
8 high visibility LED bars
Frequency: ETSI 865.6 to 867.6 MHz or FCC/IC 902 to 928 MHz

Delivery information: Includes (1) Brady FR22 Reader (B-FR22-RFID-RDR-US);(1) BFA Antenna (B-BFA-AnT-915).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty