Would you like to log in again?
Diluent C (with rAlbumin)
Supplier: New England Biolabs
B8534S
77322-306EA 37 USD InStock
77322-306
Diluent C (with rAlbumin)
Nucleic Acid Reagents Restriction Enzymes
Diluent C (with rAlbumin) is recommended for making dilutions of New England Biolabs, Inc. restriction enzymes.
  • Contains 4×1.25 ml vials
  • Restriction enzymes should be stored at their selling concentrations
  • Contains rAlbumin
Diluent C (with rAlbumin) is recommended for making dilutions of some NEB restriction enzymes. When necessary, we recommend diluting enzymes just prior to use and suggest that the final concentration of diluted enzymes be at least 1000 units/ml. Restriction enzymes should be stored at their selling concentrations. Each vial contains 1.25 ml of diluent buffer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty