Would you like to log in again?
Reusable Filter Animal Cage Covers, Tecniplast®
Supplier: Tecniplast
1290D400SU 1264C400SU 1284L400SU 1144B400SU 1144B400SU 1264C400SU 2000P400SU 2150E400SU 2154F400SU 2154F400SU
77322-368EA 90.72 USD InStock
77322-368 77322-412 10715-415 77322-420 10715-403 10715-356 10715-345 10715-323 77322-380 77322-430
Reusable Filter Animal Cage Covers, Tecniplast®
Animal Supplies Animal Cages
Securely fit Tecniplast® plastic animal cages.
  • Filter covers permit atmospheric gas passage with minimal buildup of temperature and humidity
  • Covers are constructed of transparent polysulfone with a stainless steel filter retainer frame
  • Covers fit over water bottle
Replacement filters are ready-to-use and presized to fit neatly inside the cover.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty