Would you like to log in again?
Reusable Plastic Animal Cages, Tecniplast®
Supplier: Tecniplast
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2150E00SU 2000P001 1144B00SU 2150E001 1291H001 1264C00139 1144B00SU 1284L001 1284L001 1144B001 1144B00139 1264C001 2154F001 2000P00SU 1354G001 2154F00SU 1264C00SU 2150E001 1284L051 1144B001 1264C00139 1144B00139 2000P00130 1291H00SU 1264C001 1264C00SU
77322-356EA 88.36 USD InStock
77322-356 77322-398 77322-416 10712-320 77322-370 77322-396 77322-374 77322-394 77322-372 77322-408 77322-406 10712-334 10712-318 77322-422 77322-404 10712-234 10710-022 10710-021 77322-424 10712-110 10712-154 77322-382 77322-360 77322-364 77322-362 77322-384
Reusable Plastic Animal Cages, Tecniplast®
Animal Supplies Animal Cages
Plastic animal cages provide warm surfaces ideal for housing laboratory rodents and are convenient to use and maintain. Cage bodies are designed with corner lugs (side-arms) for stackable storage without scratching or jamming. Autoclavable cage bodies are available:
  • Clear standard polycarbonate
  • Clear polycarbonate with grommet for access to sipper tube on rack
  • Clear polycarbonate with cut-away bottoms for removing waste products
  • High-temperature clear polysulfone for repeated autoclaving and better chemical resistance
  • polyetherimide for maintaining superior strength and transparency after 1000 hours of autoclaving at 134 °C (273 °F)
For a complete integrated cage system for housing laboratory rodents that meets NRC guidelines, use cages with mix and match components.
For stainless steel wire cage lids and feeders, see 10713-066 series.
For raised wire floors, see 10714-025 series. For filter covers, see 10715-120 and -321 series.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty