Would you like to log in again?
Cage Racks, Tecniplast®
Supplier: Tecniplast
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2F25B1000 2D24B1011 2F20B1000 2C30B1011 2P15B1000 2E30B1021
10716-312EA 8695.53 USD InStock
10716-312 77322-432 10716-315 10716-310 95057-890 95057-884
Cage Racks, Tecniplast®
Animal Supplies Animal Cages
Cage racks equipped with automatic watering equipment feature a Reverse-S manifold piping manufactured from corrosion-resistant stainless steel. Design permits excellent viewing of all cages (not included) and occupants, and improves hygiene, safety, and ergonomics.
  • Designed to allow positive air venting and complete water exchange in the manifold when flushing
  • Entire manifold assembly can be easily dismantled
  • Suspension-style racks minimize movement of individual cages when others are serviced or entire rack is moved, also eliminate bumping of adjacent filter covers
  • Constructed of stainless steel with nylon support rails
Shelf-style cage racks feature solid sheet stainless steel shelves for rigid, long-lasting performance.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty