Would you like to log in again?
AccuForge® Hobby Blade #11, Stainless Steel
Supplier: AccuTec Blades
AccuForge®
AGBL-2001-0000
77322-444CS 631.23 USD InStock
77322-444
AccuForge® Hobby Blade #11, Stainless Steel
Razors and Blades
AccuForge® #11 Hobby blades are constructed from stainless steel for increased sharpness and corrosion resistance. Precision acute point for fine cuts.
  • Fits all standard #1 Handles
  • High-quality stainless steel blade
  • Bulk pack of 1000 blades
  • Made in USA
Excellent for precision cutting and trimming.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty