Would you like to log in again?
AccuForge® Double Edge Blades, Unwrapped
Supplier: AccuTec Blades
AccuForge®
AGBL-6002-0000 AGBL-6005-0000
77322-446CS 8770.77 USD InStock
77322-446 77322-450
AccuForge® Double Edge Blades, Unwrapped
Razors and Blades
AccuForge® Double Edge blades feature a 3-Facet edge for exacting, clean cuts. The proprietary MicroCoat® Enrichment Technology reduces cut force and helps the blade maintain a sharp, effective edge over multiple uses.
  • Blade edge with MicroCoat® technology enhances durability and sharpness
  • 3-Facet edge for increased sharpness
  • Available degreased for reduced oil and particulates
  • Blades are supplied on a bayonet
  • Bulk unwrapped blades are ideal for high volume uses
  • Made in USA
MicroCoat® blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp and effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty