Would you like to log in again?
AccuForge® Double Edge Blades, Individually Wrapped
Supplier: AccuTec Blades
AccuForge®
AGBL-6003-0000 AGBL-6006-0000
77322-448CS 180.14 USD InStock
77322-448 77322-452
AccuForge® Double Edge Blades, Individually Wrapped
Razors and Blades
AccuForge® Double Edge blades feature a 3-Facet edge for exacting, clean cuts.
  • 3-Facet blade edge for increased sharpness
  • High-quality stainless steel
  • Available degreased for reduced oil and particulates
  • Made in USA
Blades are individually wrapped for safety.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty