Would you like to log in again?
AccuForge® Single Edge Long Blades
Supplier: AccuTec Blades
AccuForge®
AGBL-7003-0000 AGBL-7002-0000 AGBL-7001-0000 AGBL-7004-0000 AGBL-7000-0000
77322-456CS 423.59 USD InStock
77322-456 77322-464 77322-458 77322-462 77322-460
AccuForge® Single Edge Long Blades
Razors and Blades
AccuForge® Single Edge long blades feature 3-Facet edges for precise cuts. The proprietary MicroCoat® Enrichment Technology reduces cut force and helps the blade maintain a sharp, effective edge over multiple uses.
  • 3-Facet edge for increased sharpness
  • 2.25" length for extra cutting width
  • Constructed from high-quality stainless steel
  • Available in bulk pack or individually wrapped
  • Available degreased for reduced oil and particulates
  • Available unbacked
  • Made in USA
MicroCoat® blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp and effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty