Would you like to log in again?
AccuForge® GEM® Single Edge Blade Cartridge, Coated
Supplier: AccuTec Blades
AccuForge®
AGBL-7013-BULK AGBL-7013-0000
77322-476CS 2790.7 USD InStock
77322-476 77322-474
AccuForge® GEM® Single Edge Blade Cartridge, Coated
Razors and Blades
AccuForge® Single Edge blades feature 3-facet edges for sharpness. The proprietary MicroCoat® Enrichment Technology reduces cut force and increases durability.
  • Degreased blades for reduced oil and particulates
  • Cartridge inserts into reloadable safety dispenser
  • Paper-free packaging is cleanroom ready
  • Individually packed in clamshell with product key and instructions for use
  • Available in bulk pack without clamshells
  • Each cartridge contains 100 blades
  • Made in USA
MicroCoat® blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp and effective edge over multiple uses.
Use with GEM RUNNER™ Dispenser for improved safety and productivity.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty