Would you like to log in again?
AccuForge® Single Edge Blades, 3 Facet
Supplier: AccuTec Blades
AccuForge®
AGBL-7020-0000
77322-480CS 2392.03 USD InStock
77322-480
AccuForge® Single Edge Blades, 3 Facet
Razors and Blades
AccuForge® Single Edge blades feature precision ground edges for sharpness and durability. Blades are wrapped in protective paper.
  • 3-Facet edge for increased sharpness
  • High-quality stainless steel blade
  • Blades are individually wrapped for safety
  • Reclosable clamshell for safe and convenient blade storage
  • Each clamshell contains 100 blades
  • Made in USA
MicroCoat® blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp and effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty