Would you like to log in again?
AccuForge® Single Edge Blades, 10 Blade Safety Dispenser
Supplier: AccuTec Blades
AccuForge®
AGBL-7023-0000 AGBL-7035-0000 AGBL-7040-0000
77322-498CS 1794.02 USD InStock
77322-498 77322-492 77322-482
AccuForge® Single Edge Blades, 10 Blade Safety Dispenser
Razors and Blades
AccuForge® Single Edge blades feature precision ground edges for sharpness and durability.
  • Transparent safety dispenser with used blade vault
  • Choose 3-facet blades for increased sharpness, or 2-facet blades for increased durability
  • Each dispenser contains 10 blades
Each dispenser holds 10 blades and includes used blade storage.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty