Would you like to log in again?
AccuForge® Single Edge Blades, Unwrapped
Supplier: AccuTec Blades
AccuForge®
AGBL-7038-0000 AGBL-7005-0000 AGBL-7025-0000 AGBL-7047-0000 AGBL-7041-0000
77322-466CS 573.09 USD InStock
77322-466 77322-500 76780-338 77322-506 77322-496
AccuForge® Single Edge Blades, Unwrapped
Razors and Blades
AccuForge® Single Edge blades feature precision ground edges for sharpness and durability.
  • Bulk, unwrapped blades are ideal for high volume use
  • Choose 3-facet blades for increased sharpness, or 2-facet blades for increased durability
  • Available in 0.012" heavy-duty carbon steel
  • Available in No-Notch design
These AccuForge® blades are unwrapped for high volume use.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty