Would you like to log in again?
AccuForge® GEM® Single Edge Blade Cartridges, Uncoated
Supplier: AccuTec Blades
AccuForge®
AGBL-7052-0000 AGBL-7012-0000 AGBL-7014-0000 AGBL-7011-0000 AGBL-7051-0000 AGBL-7051-BULK
77322-510CS 431.9 USD InStock
77322-510 77322-478 77322-514 77322-512 77322-470 77322-472
AccuForge® GEM® Single Edge Blade Cartridges, Uncoated
Razors and Blades
AccuForge® Single Edge blades feature precision ground edges for sharpness and durability.
  • Paper-free packaging is cleanroom ready
  • Choose 3-facet blades for increased sharpness, or 2-facet blades for increased durability
  • Individually packed in clamshell with product key and instructions for use
  • Available in bulk pack without clamshells
  • Available in 0.012" heavy-duty carbon steel
  • Each cartridge contains 90 to 100 blades
Use with GEM RUNNER™ Dispenser for improved safety and productivity.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty