Would you like to log in again?
AccuForge® Contour Blades
Supplier: AccuTec Blades
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AccuForge®
AGBL-9000-0000 AGBL-9001-0000
77322-518CS 963.46 USD InStock
77322-518 77322-516
AccuForge® Contour Blades
Razors and Blades
AccuForge® contour blades feature precision ground edges for high accuracy cutting applications.
  • Great for precision cuts
  • Degreased to remove oil residue
  • Available in standard or long point
  • High-quality stainless steel
  • Made in USA
Slot-in blade allows attachment to #3 handles.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty