Would you like to log in again?
Men's Flame-Resistant/Chemical Splash Protection Lab Coats
Supplier: Bulwark Protection
KNR6RB-XL-0R KNR6RB-5L-0R KNR6RB-LG-0R KNR6RB-3L-0R KNR6RB-MD-0R KNR6RB-SM-0R KNR6RB-4L-0R KNR6RB-2L-0R
77322-576EA 240.99 USD InStock
77322-576 77322-574 77322-578 77322-580 77322-572 77322-584 77322-570 77322-582
Men's Flame-Resistant/Chemical Splash Protection Lab Coats
Workwear
This environment of ingenuity and innovation demands reliable lab PPE that provides the ultimate in protection and comfort, for all-day wear. That’s why Bulwark CP offers breakthrough safety wear designed specifically for the lab.
  • Black lay down collar for easy recognition as a CP lab coat
  • Concealed snap front closure
  • Splash-resistant pass-throughs
  • FR only knit cuffs, safer than typical open cuff
  • Double needle top-stitched chest pocket with pencil stall
  • Two double needle top-stitched lower pockets with flaps
  • Back vent for ease of walking and sitting
Regular clothing or lab coats simply won’t keep lab workers protected in the case of a thermal incident or inadvertent chemical splash, but these single and multi-hazard lab coats help provide protection that fits well and is easy to wear.
Certifications: NFPA 2112 compliant.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty