Would you like to log in again?
VWR® Disposable Cleanroom Socks
Supplier: Avantor
VWR®
CRS-D
77322-586CS 566.83 USD InStock
77322-586
VWR® Disposable Cleanroom Socks
Disposable Garments
VWR® Disposable cleanroom socks are designed to provide all day comfort inside a cleanroom. They are made from materials that prevent any particle generation while in use.
  • Single-use disposable socks
  • Poly/Nylon blend
  • Comfortable for extended use
  • Cleanroom compatible
  • Annex 1 compliant
Cleanroom socks provide comfort and durability for limited and extended wear in cleanroom environments. Suitable for use in ISO 4 to 6 (Class 10 to 1000) cleanrooms. Available in either launderable or disposable options.
Packaging: 400 pairs per case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty