Would you like to log in again?
HyFlex® 11-581 Industrial Gloves
Supplier: Ansell
HyFlex®
11581120 11581100 11581080 11581070 11581090 11581060 11581110
77322-774CS 1680 USD InStock
77322-774 77322-764 77322-772 77322-762 77322-768 77322-766 77322-770
HyFlex® 11-581 Industrial Gloves
Gloves
HyFlex® 11-581 industrial gloves feature an ultra-lightweight design, delivering 20% lighter EN ISO Cut F and ANSI/ISEA 105-2024 Cut A6 protection.
  • Ultra-lightweight design for unparalleled comfort and dexterity
  • 6 times greater cut resistance
  • Ideal for dry and slightly oily applications
  • Palm dipped, thumbcrotch reinforcement
  • Silicone-free and anti-static
HyFlex® 11-581 industrial gloves offer 6 times greater cut resistance thanks to the Next Generation single HPPE yarn developed with INTERCEPT™ Cut Resistance Technology for reliable hand protection. Featuring FORTIX™ Technology and thumb crotch reinforcement, these gloves ensure safer handling in abrasive conditions. In addition, HyFlex® 11-581 protective gloves allow wearers to work with multiple touchscreen devices while keeping them on.
HyFlex® 11-581 industrial gloves are recommended for use in automotive, machinery, equipment, metal fabrication, and aerospace industries.
Certifications: ANSI ABR 4. ANSI Cut A6. Dermatologically Tested. EN 388:2016 +A1:2018, EN 16350:2014, EN ISO 21420:2020. Food Contact approved. OEKO-TEX® Hohenstein.
Packaging: Gloves are packaged 12 pairs per bag, 12 bags/carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty