Would you like to log in again?
HyFlex® 11-591 Industrial Gloves, Vendor Pack
Supplier: Ansell
HyFlex®
11591060-VEND 11591100-VEND 11591120-VEND 11591080-VEND 11591090-VEND 11591110-VEND 11591070-VEND
77322-822CS 1944.96 USD InStock
77322-822 77322-820 77322-826 77322-816 77322-824 77322-814 77322-818
HyFlex® 11-591 Industrial Gloves, Vendor Pack
Gloves
HyFlex® 11-591 industrial gloves provide enhanced comfort, with a 20% lighter design that allows extended use on varied tasks.
  • Ultra-lightweight design
  • 12 times greater cut resistance for reliable hand protection
  • Palm dipped, thumbcrotch reinforcement
  • Silicone-free and anti-static
HyFlex® 11-591 industrial gloves offer 12 times greater cut resistance thanks to the Next Generation single HPPE yarn developed with INTERCEPT™ Cut Resistance Technology for reliable hand protection. Featuring a 20% lighter design, these gloves provide enhanced comfort, and allow workers to perform varied tasks while keeping hands protected throughout extended use. Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting handling in abrasive conditions. EN 407 contact heat level 1 to protect hands while handling hot parts.
HyFlex® 11-591 industrial gloves are recommended for use in automotive, glass manufacturing, heavy manufacturing, metal fabrication, machinery, equipment, and pulp/paper industries.
Certifications: ANSI Cut A9. ANSI ABR 5. EN 388:2016 +A1:2018, EN407:2020, EN ISO 21420:2020. Food Contact approved. Category II.
Packaging: Gloves are packaged 12 pairs per paper band bundle, 12 paper band bundles per carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty