Would you like to log in again?
AlphaTec® 38-001 Butyl Gloves
Supplier: Ansell
AlphaTec®
38001PP100 38001PP090 38001PP110 38001PP080 38001PP070
77322-830CS 1008 USD InStock
77322-830 77322-834 77322-832 77322-836 77322-828
AlphaTec® 38-001 Butyl Gloves
Gloves
AlphaTec® 38-001 butyl gloves offer superior protection against highly aggressive chemicals, including aldehydes, ketones, esters, and concentrated mineral acids.
  • Unique butyl formulation for a soft feel with a raised diamond pattern
  • Extended forearm protection
  • Excellent dry and wet grip to enhance dexterity and reduce hand fatigue
  • Wide cuff to allow donning over protection clothing
Designed for comfort, AlphaTec® 38-001 industrial gloves feature a unique butyl formulation to facilatate a soft feel, protecting hands while facilitating donning and doffing. The ultra-soft butyl formulation maximizes flexibility and dexterity, with a raised diamond pattern that enhances grip and ensures safer handling.
AlphaTec® 38-001 industrial gloves are recommended for use in automotive, life science, and chemical industries.
Certifications: EN ISO 21420:2020, EN ISO 373-1:2016 Type A, EN ISO 374-5:2016.
Packaging: Gloves are packaged 1 pair per bag, 36 bags per carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty