Would you like to log in again?
Advanced MX Precision Balances, Legal for Trade
Supplier: Mettler-Toledo
30665195 30665198 30665192 30665189
77323-122EA 11293 USD InStock
77323-122 77323-114 77323-126 77323-118
Advanced MX Precision Balances, Legal for Trade
Balances and Scales
METTLER TOLEDO Advanced MX Precision Legal for Trade Balances feature a large weighing pan, robust design, and precise weigh cell with FACT automatic internal adjustment.
  • SmartPan
  • Statistical data analysis
  • User management; Leveling assistant
  • StatusLight
  • 3 interfaces: USB-A, USB-C, Ethernet
  • Bluetooth option
  • EasyDirect software
  • Full metal housing with overload protection
The 7" intuitive touchscreen with user management provides automatic data logging, statistical analysis and easy access to 9 built-in weighing applications. Transfer data to a printer or computer for error-free documentation. Built with IP 54 protection and robust construction, these balances will deliver high quality performance for years to come. Enable the power-saving mode when balance is not in use to saves costs and reduces energy consumption.
Certifications: OIML/NTEP approved.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty