Would you like to log in again?
Analytical Balances, Advanced MR
Supplier: Mettler-Toledo
30666206 30666206 30666203 30666209 30666209 30666203
77323-136EA 5526 USD InStock
CA77323-132 77323-136 CA77323-136 77323-128 CA77323-128 77323-132
Analytical Balances, Advanced MR
Balances and Scales
METTLER TOLEDO Advanced MR Analytical Balances offer a highly precise weigh cell with FACT automatic internal adjustment.
  • Guided leveling
  • Four interfaces: USB-A, USB-C, RS232, Ethernet
  • Bluetooth option
User management within the 4.5" touchscreen increases efficiency of daily tasks with 8 built-in applications and statistical data analysis. Designed with full metal housing and overload protection, these balances will provide high quality performance for years to come. When balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty