Would you like to log in again?
Precision Balances, Legal For Trade, Standard MA
Supplier: Mettler-Toledo
30697459 30697427 30697444 30697424 30697453 30697441 30697447 30697430 30697421 30697450 30697456
77323-210EA 2467 USD InStock
77323-210 77323-222 77323-234 77323-202 77323-214 77323-226 77323-238 77323-206 77323-218 77323-230 77323-242
Precision Balances, Legal For Trade, Standard MA
Balances and Scales
METTLER TOLEDO Standard MA Legal for Trade Precision Balances are highly accurate, simple to operate and built with high quality materials for proven long lasting performance.
  • Statistical data analysis
  • Front level indicator
  • PBT top housing
  • Two interfaces: USB-A, RS232
  • Bluetooth option
The precise weigh cell and internal adjustment provide accurate results. Intuitive onscreen guidance within the LCD hybrid touchscreen allows you to easily navigate through 10 built in applications and manage weighing data. The metal base and overload protection defend the balance against rough handling in any weighing environment.
Certifications: OIML/NTEP Approved.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty