Would you like to log in again?
Precision Balances, Legal For Trade, Standard MA
Supplier: Mettler-Toledo
30697482 30697485 30697479 30697476
77323-254EA 8169 USD InStock
77323-254 77323-246 77323-258 77323-250
Precision Balances, Legal For Trade, Standard MA
Balances and Scales
METTLER TOLEDO Standard MA Legal for Trade Precision Balances have a large weighing pan, robust design and highly accurate weigh cell with internal adjustment.
  • Statistical data analysis
  • Front level indicator
  • Metal base, PBT top housing
  • Overload protection
  • Two interfaces: USB-A, RS232
  • Bluetooth option
The hybrid LCD touchscreen increases efficiency in the workflow with intuitive onscreen guidance and user management. Easily navigate 10 built-in applications and export weighing data. Built with robust construction to defend against external influences and provide high quality service for years to come. Easily export weighing data to a computer, printer, or USB storage device.
Certifications: OIML/NTEP Approved.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty