Would you like to log in again?
Precision Balances, Standard MA-P
Supplier: Mettler-Toledo
30697493 30697487 30697493 30697487 30697490 30697490
77323-268EA 1963 USD InStock
CA77323-264 77323-268 CA77323-268 77323-260 CA77323-260 77323-264
Precision Balances, Standard MA-P
Balances and Scales
METTLER TOLEDO Standard MA-P Precision Balances are simple to operate and portable for on-the-go weighing, without compromising on accuracy or performance.
  • Strain gauge weighing cell
  • Internal adjustment
  • LCD hybrid touchscreen
  • 10 built-in applications
  • Power cable and battery operated
  • Front level indicator
  • Overload protection
  • Two interfaces: USB-A, RS232
  • Bluetooth option
The precise weigh cell delivers highly accurate results with each use. Increase efficiency in the workflow with the intuitive touchscreen that guides you through each weighing procedure. Easily export data to a connected printer, computer, or USB device.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty