Would you like to log in again?
Analytical Balances, Standard LA
Supplier: Mettler-Toledo
30823718 30823719 30823720 30823718 30823719 30823720
77323-276EA 2143 USD InStock
77323-276 77323-278 CA77323-276 CA77323-278 77323-280 CA77323-280
Analytical Balances, Standard LA
Balances and Scales
METTLER TOLEDO Standard LA Analytical Balances have a precise weigh cell and external adjustment. The bright, backlit LCD display is equipped with 3 built-in applications for piece counting and dynamic weighing.
  • Front level indicator
  • ABS top housing
  • RS232 interface
  • Bluetooth option
  • Transfer weight function for easy data transfer
Designed with robust construction, the overload protection and metal base can withstand tough weighing environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty