Improve the performance, microbial barrier and cleanliness of your autoclave peel pouches.
- Manufactured and packaged in a controlled cleanroom environment
- Autoclavable at 121 to 125 °C
- Peels open cleanly
- Chevron peel
- Tear and puncture resistant
- Hydrophobic
- Lint-free
- Heat seals closed
- Integrated steam sterilization indicator
A significant upgrade over paper-based steam sterilization pouches. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do.
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
Delivery information: Certificates of Conformance sent with each shipment.