Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zebra Scan-One-Print-One Label Replicator

Supplier: Computype
Zebra
18367821
77335-368EA 3326 USD InStock
77335-368
Zebra Scan-One-Print-One Label Replicator
Label Makers
Computype laboratory label replicator systems are specially modified to solely perform the task of replicating individual labels. Simply scan the label you’d like to replicate to print a new label. Compatible with TS976 tube and vial labels.

  • Zebra ZD421 300DPI enhanced printer
  • Print resolution: 300 dpi
  • Thermal transfer printing
  • Scanner LS2208 RS232 DB9M white
  • ZBI key kit 1 printer version 2.0

Duplicate scanned barcode onto one of five label sizes (in inches) selectable from included scan menu:
76635-386 TS976, 1×0.5"
76635-390 TS976, 1×1"
76635-394 TS976, 1.5×0.75"
76635-398 TS976, 2×1"
10769-986 TS839P, 2.5×0.25"

Label format can include one line of text that can be defined with included scan menu. Text can be positioned above or below barcode. Variable quantity up to 99 copies can be selected from included scan menu.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty