Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Self-Sealing Sterilization Peel Pouches, Medical Paper/Film, with Steam Indicator

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PPSS10X14CF PPSS12X18CF PPSS035X055CF PPSS075X13CF PPSS075X13 PPSS08X16CF PPSS10X14 PPSS12X18 PPSS08X16 PPSS055X105CF PPSS035X09 PPSS055X105 PPSS035X09CF PPSS035X055
77391-778CS 119.16 USD InStock
77391-778 77391-776 77391-788 77391-796 77391-774 77391-786 77391-772 77391-794 77391-784 77391-792 77391-770 77391-782 77391-790 77391-780
Self-Sealing Sterilization Peel Pouches, Medical Paper/Film, with Steam Indicator
Bags Sterilization Pouches
Designed for steam sterilization cycles (121 to 135 °C).

  • Peels open cleanly
  • Triple weld factory seals
  • Convenient, time-saving self-seal closure system
  • Strong, rip- and tear-resistant 60 g/m² medical grade paper material
  • Good barrier characteristics prevent microbial penetration
  • Sub-packed into sealed labeled poly bags with manufacturing lot number
  • Date of manufacture on label for complete traceability

Significant improvement versus cutting and wrapping with sheets and autoclave tape.

Certifications: Certificates of Conformance sent with each shipment.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty