Would you like to log in again?
StarterKit StarterKit −30 to +70 °C Temperature RH, with AccessPoint, AssetTag and Six Credits
Supplier: SenseAnywhere
0096647495477
77391-872EA 619 USD InStock
77391-872
StarterKit StarterKit −30 to +70 °C Temperature RH, with AccessPoint, AssetTag and Six Credits
Monitoring Systems CO2 and Temperature Monitoring Systems
Experience the SenseAnywhere StarterKit −30 to +70 °C, with the AiroSensor 20-20-25 for accurate temperature and RH tracking. Includes a secure Indoor AccessPoint, innovative AssetTagging, and 6 Credits for effortless sensor management, offering up to 10 years of maintenance-free, reliable monitoring across diverse environments.
  • Accurate monitoring from −30 to +70 °C
  • 24/7 data gives you a complete audit trail
  • Access your data anywhere and anytime via the Cloud
  • Get notified via email, text or voice when limits are exceeded
Introducing the SenseAnywhere AiroSensor 20-20-25: a highly compact and robust wireless data logger designed for precise temperature, humidity and motion tracking. This user-friendly device seamlessly connects to the Cloud for data storage. Effortless to install and maintain, it boasts an ultra-low power design with up to 10 years of operation. With a wireless transmission range of up to 600 meters (line of sight) to an AccessPoint, it's perfectly suited to monitor relative humidity inside storage rooms.
Indoor AccessPoint - The SenseAnywhere Indoor AccessPoint is the cornerstone of reliable and secure data transmission. It connects our Sensors to the SenseAnywhere Cloud Service, ensuring robust, two-way, fully encrypted communication. Sensors automatically detect the AccessPoint, seamlessly transmitting data to our central database. With advanced protocols for re-transmission and duplicate packet filtering, we guarantee that no event is ever missed. Designed specifically for indoor environments, this AccessPoint is an integral part of our robust surveillance solution.
AssetTagging - AssetTagging by SenseAnywhere revolutionizes sensor calibration and monitoring continuity. This system allows for uninterrupted data logging with interchangeable loggers, eliminating gaps during sensor recalibration and the hassle of resetting alarms or reports. The AssetTag, featuring a QR code, facilitates quick and easy swapping of AiroSensors. When an AiroSensor needs recalibration, simply swap the AiroSensor without any data loss. All pre-set alarms and configurations automatically apply to the new AiroSensor, ensuring a gap-free monitoring experience. With AssetTagging, data loggers can be switched anytime, maintaining continuous and reliable surveillance.
Ordering information: Explore the full range of SenseAnywhere products and services by visiting the Documentation tab, selecting 'Additional Product Details,' and opening the 'Which SenseAnywhere Data Logger Do I Need' document.
Delivery information: Begin precise RH and temperature surveillance of your vital assets ranging from −30 to +70 °C with our comprehensive StarterKit. This kit includes an AiroSensor 20-20-25, an AccessPoint for reliable data transmission, an AssetTag for seamless sensor management, and six credits for immediate use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty