Would you like to log in again?
Rat MD's™ Baytril and Rimadyl Tablets
Supplier: Bio-Serv
F06801 F07955 F06801 F07955
77391-932EA 175.15 USD InStock
CA77391-932 77391-932 CA76285-254 76285-254
Rat MD's™ Baytril and Rimadyl Tablets
Animal Supplies Animal Feed
Baytril is an effective antibiotic drug and Rimadyl an effective anti-inflammatory drug both which are used in a variety of species including rodents.
  • Each 5 gram tablet contains: 2 mg Baytril and 2 mg Rimadyl
  • Highly palatable, nutritionally complete, grain-based, bacon or chocolate flavored tablets
  • Easy to administer: Place on cage floor for easy access
  • Safe alternative to traditional dosing methods
  • Cold tableting process eliminates drug degradation
  • Can be customized to meet specific dosage requirements
  • Nutritionally assayed
  • More convenient dosing method and minimizes risk of dehydration compared to using medicated water
MD's are highly palatable and encourage appetite post-operatively promoting a more rapid recovery versus other conventional methods. They are an ideal pain-free alternative to stressful and injurious post-operative injections.
Baytril and Rimadyl combination MD's allow for the administration of both medications easily and affordably.
Introduction of Bio-Serv's Placebo or MD tablet 1 to 2 days prior to surgery will decrease novelty and increase acceptance.
Rat average daily dose: One 5 gm tablet/rat per day provides daily medication needs and is a dietary supplement to standard feed.
Ingrediants: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty