Would you like to log in again?
SD Pro™ Primate Probiotic, Fiber Plus Tablet
Supplier: Bio-Serv
SD Pro™
PB120-100 PB120-100
77391-952PK 281.15 USD InStock
CA77391-952 77391-952
SD Pro™ Primate Probiotic, Fiber Plus Tablet
Animal Supplies Animal Feed
SD Pro™ is a series of species derived origin probiotics designed specifically for the host species that combines the benefits of Lactobacillus reuteri and Saccharomyces boulardii.
  • SD Pro™ tablets contains live culture, which is sensitive to heat and moisture
  • Highly palatable, sweet cherry flavored tablets
L. reuteri is a natural bacterial inhabitant of a healthy GI tract and contains antimicrobial properties. S. boulardii regulates intestinal microbial homeostasis and prevents the colonization of pathogens within the GI tract.
Each tablet contains ~1.25 billion CFU of Lactobacillus reuteri, ~140 mg Saccharomyces boulardii and 300 mg L-Glutaminein a highly palatable, easy to dose flavored tablet.
Fiber Plus includes the added benefits of 1 gm of soluble fiber (Inulin) and 1 gm of insoluble fiber (Cellulose) to aid in gastrointestinal support.
Ingredients: Sucrose, dextrose, cellulose, inulin, L-glutamine, flavoring, yeast culture, hydrolyzed yeast, calcium silicate, magnesium stearate, dried Lactobacillus reuteri, fermentation product, soybean oil, sodium aluminosilicate
Note: If non-human primate is on antibiotic therapy, administer SD Pro 4 to 6 hours after antibiotic dosage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty