Would you like to log in again?
SD Pro™ Mini, Primate Probiotic Tablets
Supplier: Bio-Serv
SD Pro™
PB130-100 PB130-100
77391-954EA 222.58 USD InStock
CA77391-954 77391-954
SD Pro™ Mini, Primate Probiotic Tablets
Animal Supplies Animal Feed
SD Pro™ is a series of species derived origin probiotics designed specifically for the host species that combines the benefits of Lactobacillus reuteri and Saccharomyces boulardii.
  • L. reuteri is a natural bacterial inhabitant of a healthy GI tract and contains antimicrobial properties
  • S. boulardii regulates intestinal microbial homeostasis and prevents the colonization of pathogens within the GI tract
  • Each tablet contains ~0.4 billion CFU of Lactobacillus reuteri, ~35 mg Saccharomyces boulardii and 75 mg L-Glutaminein a highly palatable, easy to dose flavored tablet
  • Highly palatable, pina colada flavored tablets
  • Shelf life: 6 months from date of manufacture - refrigerated
SD Pro tablets contains live culture, which is sensitive to heat and moisture.
Ingredients: Sucrose, dextrose, L-glutamine, flavoring, yeast culture, hydrolyzed yeast, calcium silicate, magnesium stearate, dried lactobacillus reuteri, fermentation product, soybean oil, sodium aluminosilicate.
Feeding guidelines: One tablet treats an average 400 gm marmoset. Note: If non-human primate is on antibiotic therapy, administer SD Pro 4 to 6 hours after antibiotic dosage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty