Would you like to log in again?
SD Pro™ Mini Primate Probiotic, Magic Tabs
Supplier: Bio-Serv
SD Pro™
PB135-100 PB135-100
77391-956EA 433.43 USD InStock
CA77391-956 77391-956
SD Pro™ Mini Primate Probiotic, Magic Tabs
Animal Supplies Animal Feed
SD Pro magic tabs are a therapetutic cocktail of beneficial probiotics, potassium gluconate and bismuth subsalicylate for use in primates with idiopathic diarrhea. Coined 'Magic Tabs' as they are an all-in-one tasty tablet conferring similar benefits as bananas, Pepto Bismol and microbiome support.
  • Highly palatable, sweet pina colada flavored tablets
  • Shelf life: 6 months from date of manufacture - refrigerated
  • SD Pro tablets contains live culture, which is sensitive to heat and moisture
SD Pro™ is a series of species derived origin probiotics designed specifically for the host species that combines the benefits of Lactobacillus reuteri and Saccharomyces boulardii. S. boulardii regulates intestinal microbial homeostasis and prevents the colonization of pathogens within the GI tract. L. reuteri is a natural bacterial inhabitant of a healthy GI tract and contains antimicrobial properties. Each tablet contains ~0.4 billion CFU of Lactobacillus reuteri , 42.5 mg Saccharomyces boulardii, 0.65 mEq K+, 36 mg Bismuth Subsalicylate and 75 mg L-Glutamine in a highly palatable, easy to dose flavored tablet.
Ingredients: Sucrose, dextrose, L-glutamine, berry flavoring, renal K, L-glutamine, yeast culture, hydrolyzed yeast, calcium silicate, magnesium stearate, dried lactobacillus reuteri, fermentation product, soybean oil, bismuth subsalicylate, sodium aluminosilicate.
Feeding guidelines: One tablet treats an average 400 gm marmoset. Note: If non-human primate is on antibiotic therapy, administer SD Pro 4 to 6 hours after antibiotic dosage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty