Would you like to log in again?
Mini Mouse Huts, Certified
Supplier: Bio-Serv
S3450 K3450 S3450 K3450
77391-958CS 339.43 USD InStock
CA77391-958 77391-958 CA77391-944 77391-944
Mini Mouse Huts, Certified
Animal Supplies Animal Cages
Provides sheltering and nesting opportunities.
  • Made of non-recycled, 100% virgin wood pulp
  • Low profile - 2" tall fits in most shoebox cages
  • Economical and disposable enrichment
  • Acrylamide free
  • Autoclavable
Certifications: Certified (contaminant screened).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty