Would you like to log in again?
Sentino® Filter Dispenser, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sentino®
68121ME 13184 13184 68121ME
77391-988PK 643.15 USD InStock
CA10147-394 CA77391-988 77391-988 10147-394
Sentino® Filter Dispenser, Cytiva (Formerly Pall Lab)
Membrane Dispensers
Filter dispensers are ideal for high throughput microbiology testing laboratories looking for more ergonomic, faster ways to handle individual membranes. The Sentino® filter dispenser is designed to aseptically dispense one disc of membrane at the touch of a button.
  • Easy to load without jamming. Guide plates assist loading and keep refill web in alignment during use
  • Soft touch keypad for aseptically dispensing membrane discs
  • Designed to hold 200 membrane discs
  • Compact, portable design with miniaturised refill pack
  • Use plugged into main power source or charge battery for use anywhere. Lights indicate when battery charge is running low
  • Easy to clean
The Sentino® filter dispenser can be used for: Drinking and wastewater analysis, environmental and recreational water monitoring, personal care product manufacturing, beverage monitoring, pharmaceutical and industrial water system monitoring, pharmaceutical raw materials, intermediates and final product.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty