Would you like to log in again?
Capto™ Core 400 Multimodal Chromatography Resins
Supplier: Cytiva
Capto™
17372402 17372401 17372401 17372402
77392-080EA 966.51 USD InStock
CA77392-080 77392-080 77392-078 CA77392-078
Capto™ Core 400 Multimodal Chromatography Resins
Protein Purification Protein Purification Resins
Capto Core 400 is designed for intermediate purification and polishing of viruses and other large biomolecules.
  • Significantly improved productivity enabled by up to 100‑fold higher sample load and significantly higher flow rates compared with SEC
  • Bead with ligand‑activated core and inactive shell allows efficient capture of impurities, while target molecules are collected in the flowthrough
  • Straightforward optimization due to flow-through chromatography and robust performance, allowing for a wide window of operation
Novel core bead technology and multimodal ligand give Capto Core 400 dual functionality, size exclusion and binding chromatography, in one chromatography resin.
Capto Core 400 is an excellent alternative to size‑exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacture.
Chemical stability: All commonly used aqueous buffers, 1 M sodium hydroxide (NaOH), 6 M guanidine hydrochloride, 30% isopropanol, and 70% ethanol.
Caution: Avoid using oxidizing agents, citrate buffers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty