Would you like to log in again?
Thermo Scientific™ Nunc™ EASYFlask™ with the Nunclon™ Supra Surface Treatment
Supplier: Thermo Fisher Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Thermo Scientific™ Nunc™
156380 156372 156376 156378 156374 156382
77392-110CS 521.16 USD InStock
77392-110 77392-108 77392-116 77392-118 77392-112 77392-114
Thermo Scientific™ Nunc™ EASYFlask™ with the Nunclon™ Supra Surface Treatment
Flasks Cell Culture Flasks
Experience optimal performance with Thermo Scientific Nunc™ EASYFlasks™ with the Nunclon™ supra surface treatment for culturing a wide variety of fastidious adherent cells.
  • Consistent lot-to-lot performance
  • Sterile with SAL 10⁻⁶
  • Non-cytotoxic
  • Non-pyrogenic
  • Biocompatible per USP Class VI for raw materials
  • Shelf life five years
The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon™ supra surface is more hydrophilic than the Nunclon™ delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Nunclon™ supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Thus, Nunclon™ supra cultureware is a great xeno-free solution for cell therapy research.
Packaging: 25 cm² models: 10 flasks per pack; others: 5 flasks per pack.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty